Merck
  • Hereditary thrombophilia: identification of nonsense and missense mutations in the protein C gene.
Hereditary thrombophilia: identification of nonsense and missense mutations in the protein C gene.
Proceedings of the National Academy of Sciences of the United States of America (1987-05-01)
G Romeo, H J Hassan, S Staempfli, L Roncuzzi, L Cianetti, A Leonardi, V Vicente, P M Mannucci, R Bertina, C Peschle
ABSTRACT
The structure of the gene for protein C, an anticoagulant serine protease, was analyzed in 29 unrelated patients with hereditary thrombophilia and protein C deficiency. Gene deletion(s) or gross rearrangement(s) was not demonstrable by Southern blot hybridization to cDNA probes. However, two unrelated patients showed a variant restriction pattern after Pvu II or BamHI digestion, due to mutations in the last exon: analysis of their pedigrees, including three or seven heterozygotes, respectively, with approximately 50% reduction of both enzymatic and antigen level, showed the abnormal restriction pattern in all heterozygous individuals, but not in normal relatives. Cloning of protein C gene and sequencing of the last exon allowed us to identify a nonsense and a missense mutation, respectively. In the first case, codon 306 (CGA, arginine) is mutated to an inframe stop codon, thus generating a new Pvu II recognition site. In the second case, a missense mutation in the BamHI palindrome (GGATCC----GCATCC) leads to substitution of a key amino acid (a tryptophan to cysteine substitution at position 402), invariantly conserved in eukaryotic serine proteases. These point mutations may explain the protein C-deficiency phenotype of heterozygotes in the two pedigrees.