Merck
  • Phosphorylation of Hsp20 Promotes Fibrotic Remodeling and Heart Failure.
Phosphorylation of Hsp20 Promotes Fibrotic Remodeling and Heart Failure.
JACC. Basic to translational science (2019-05-08)
George T Gardner, Joshua G Travers, Jiang Qian, Guan-Sheng Liu, Kobra Haghighi, Nathan Robbins, Min Jiang, Yutian Li, Guo-Chang Fan, Jack Rubinstein, Burns C Blaxall, Evangelia G Kranias
ABSTRACT
Cardiomyocyte-specific increases in phosphorylated Hsp20 (S16D-Hsp20) to levels similar to those observed in human failing hearts are associated with early fibrotic remodeling and depressed left ventricular function, symptoms which progress to heart failure and early death. The underlying mechanisms appear to involve translocation of phosphorylated Hsp20 to the nucleus and upregulation of interleukin (IL)-6, which subsequently activates cardiac fibroblasts in a paracrine fashion through transcription factor STAT3 signaling. Accordingly, treatment of S16D-Hsp20 mice with a rat anti-mouse IL-6 receptor monoclonal antibody (MR16-1) attenuated interstitial fibrosis and preserved cardiac function. These findings suggest that phosphorylated Hsp20 may be a potential therapeutic target in heart failure.
Sigma-Aldrich
MTT Cell Growth Assay Kit, MTT Cell Growth Assay is a colorimetric assay that can be used for either proliferation or complement-mediated cytotoxicity assays.