Lutein
Synonym(s):
Xanthophyll, α-Carotene-3,3′-diol, β, ε-carotene, β, ε-carotene-3,3′-diol, Lutein, β,ε-Carotene-3,3′-diol, Xanthophyll solution
Empirical Formula (Hill Notation):
C40H56O2
CAS Number:
Molecular Weight:
568.87
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique