
Tretinoin

Synonym(s):
Retinoic acid, ATRA, Tretinoin, Vitamin A acid, all-trans-Retinoic acid
Empirical Formula (Hill Notation):
C20H28O2
CAS Number:
Molecular Weight:
300.44
Products
Product Category
Biological Source
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3