Skip to Content
Merck
D-Camphor certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

D-Camphor

D-Camphor certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
2-Bornanone, 2-Camphanone, D-Camphor
Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23

Products

Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Agency
Format
Quality Segment
Application
Optical Isomer
API Family
Reference Material Type
Special Grade
Technique