Skip to Content
Merck
vibrant-m

Vinorelbine tartrate

vibrant-m
Synonym(s):
Vinorelbine ditartrate salt
Empirical Formula (Hill Notation):
C45H54N4O8 · 2C4H6O6
CAS Number:
Molecular Weight:
1079.11

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade