Skip to Content
CZ
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Titanium(IV) oxide, mixture of rutile and anatase
Synonym(s):
Titanium dioxide
Linear Formula:
TiO
2
CAS Number:
13463-67-7
Molecular Weight:
79.87
Products
Brand
Sigma-Aldrich (21)
Physical Form
Quality Segment
Filter Products
Showing 1-20 of 21
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
700347
Titanium(IV) oxide, mixture of rutile and anatase,
nanoparticles, <150 nm particle size (volume distribution, DLS), dispersion, 40 wt. % in H
2
O, 99.5% trace metals basis
Expand
View Pricing
913928
Titanium(IV) oxide, mixture of rutile and anatase,
dispersion, 10 wt. % in water, 150 nm particle size
Expand
View Pricing
914576
Titanium(IV) oxide, mixture of rutile and anatase,
150 nm particle size, dispersion, 5 wt. % in water
Expand
View Pricing
913553
Titanium(IV) oxide, mixture of rutile and anatase,
300 nm particle size, dispersion, 5 wt. % in water
Expand
View Pricing
914673
Titanium(IV) oxide, mixture of rutile and anatase,
dispersion, 10 wt. % in ethanol, 150 nm particle size
Expand
View Pricing
914835
Titanium(IV) oxide, mixture of rutile and anatase,
dispersion, 5 wt. % in ethanol, 150 nm particle size
Expand
View Pricing
914320
Titanium(IV) oxide, mixture of rutile and anatase,
dispersion, 5 wt. % in water, 500 nm particle size
Expand
View Pricing
914703
Titanium(IV) oxide, mixture of rutile and anatase,
dispersion, 10 wt. % in water, 900 nm particle size
Expand
View Pricing
914177
Titanium(IV) oxide, mixture of rutile and anatase,
dispersion, 10 wt. % in water, 300 nm particle size
Expand
View Pricing
913820
Titanium(IV) oxide, mixture of rutile and anatase,
dispersion, 5 wt. % in ethanol, 500 nm particle size
Expand
View Pricing
914606
Titanium(IV) oxide, mixture of rutile and anatase,
900 nm particle size, dispersion, 5 wt. % in water
Expand
View Pricing
914193
Titanium(IV) oxide, mixture of rutile and anatase,
dispersion, 10 wt. % in water, 500 nm particle size
Expand
View Pricing
914061
Titanium(IV) oxide, mixture of rutile and anatase,
dispersion, 5 wt. % in water, 700 nm particle size
Expand
View Pricing
914932
Titanium(IV) oxide, mixture of rutile and anatase,
300 nm particle size, dispersion, 10 wt. % in ethanol
Expand
View Pricing
914452
Titanium(IV) oxide, mixture of rutile and anatase,
dispersion, 10 wt. % in water, 700 nm particle size
Expand
View Pricing
913677
Titanium(IV) oxide, mixture of rutile and anatase,
dispersion, 10 wt. % in ethanol, 500 nm particle size
Expand
View Pricing
913561
Titanium(IV) oxide, mixture of rutile and anatase,
dispersion, 5 wt. % in ethanol, 300 nm particle size
Expand
View Pricing
914118
Titanium(IV) oxide, mixture of rutile and anatase,
dispersion, 5 wt. % in ethanol, 700 nm particle size
Expand
View Pricing
913952
Titanium(IV) oxide, mixture of rutile and anatase,
dispersion, 10 wt. % in ethanol, 700 nm particle size
Expand
View Pricing
914355
Titanium(IV) oxide, mixture of rutile and anatase,
dispersion, 5 wt. % in ethanol, 900 nm particle size
Expand
View Pricing
Page 1 of 2
Page 1 of 2