Skip to Content
CZ
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ammonium acetate solution
Linear Formula:
CH
3
COONH
4
CAS Number:
631-61-8
Molecular Weight:
77.08
Products
Product Category
buffers (3)
Brand
Sigma-Aldrich (3)
Feature
Formula Weight
pH Value
Physical Form
Product Line
Quality Segment
Application
Markush Class
Markush Group
Sterilization Method
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
A2706
Ammonium acetate solution,
for molecular biology, 7.5 M
Expand
View Pricing
09691
Ammonium acetate solution,
BioUltra, for molecular biology, ~5 M in H
2
O
Expand
View Pricing
SRE0035
Ammonium acetate solution,
Expand
View Pricing