Skip to Content
Merck
Iron(III) chloride solution 0.2 M in 2-methyltetrahydrofuran

Iron(III) chloride solution

Iron(III) chloride solution 0.2 M in 2-methyltetrahydrofuran
Synonym(s):
Ferric chloride solution
Linear Formula:
FeCl3
CAS Number:
Molecular Weight:
162.20

Products

Product Category
Brand
Sustainability
Feature
Formula Weight
Physical Form
Quality Segment
Reagent Type
Core
Special Grade
Showing 1-2 of 2