Skip to Content
CZ
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Gentamicin solution
CAS Number:
1405-41-0
Products
Product Category
antibiotics (3)
Brand
Sigma-Aldrich (3)
Biological Source
Color
Physical Form
Product Line
Activity Spectrum
Quality Segment
Mode of Action
Sterilization Method
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
G1397
Gentamicin solution,
50 mg/mL in deionized water, liquid, 0.1 μm filtered, BioReagent, suitable for cell culture
Expand
View Pricing
G1272
Gentamicin solution,
10 mg/mL in deionized water, liquid, 0.1 μm filtered, BioReagent, suitable for cell culture
Expand
View Pricing
G1522
Gentamicin solution,
Hybri-Max
™
, 0.1 μm filtered, 50 mg/mL gentamicin, BioReagent, suitable for hybridoma
Expand
View Pricing