Skip to Content
Merck
Sepharose™ CL-2B Cross-linked

Sepharose CL-2B

Sepharose™ CL-2B Cross-linked
CAS Number:

Products

Brand
Physical Form
Quality Segment
Particle Size (µm)
Showing 1-1 of 1
Compare
Product Number
Description
Pricing