Skip to Content
Merck
Sepharose™ 6B Cytiva 17-0110-01, pack of 1 L

Sepharose 4B

Sepharose™ 6B Cytiva 17-0110-01, pack of 1 L
Synonym(s):
Agarose, Beaded
CAS Number:

Products

Product Category
Brand
Biological Source
Feature
Manufacturer
Physical Form
Quality Segment
Package Size
Particle Size (µm)
Technique