
Dyclonine hydrochloride

Synonym(s):
1-(4-Butoxyphenyl)-3-(1-piperidinyl)-1-propanone
Empirical Formula (Hill Notation):
C18H27NO2 · HCl
CAS Number:
Molecular Weight:
325.87
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing