C1 Esterase Inhibitor from human plasma
Synonym(s):
Complement C1 esterase inhibitor, Esterase inhibitor C-1 from human plasma
CAS Number:
Products
Product Category
Brand
Biological Source
Gene Alias
Physical Form
Purity
Tissue
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing