Skip to Content
CZ
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ascentis
®
Express 90 Å ES-C18, 2.0 µm HPLC Column
Products
Brand
Supelco (6)
Agency
USP Suitability
ID (mm)
Length (cm)
Quality Segment
Package Size
Particle Size (µm)
Pore Size (Å)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
50609-U
Ascentis
®
Express 90 Å ES-C18, 2.0 µm HPLC Column,
column L × I.D. 10 cm × 2.1 mm, 2.0 μm particle size
Expand
View Pricing
50610-U
Ascentis
®
Express 90 Å ES-C18, 2.0 µm HPLC Column,
column L × I.D. 15 cm × 2.1 mm, 2.0 μm particle size
Expand
View Pricing
50608-U
Ascentis
®
Express 90 Å ES-C18, 2.0 µm HPLC Column,
column L × I.D. 5 cm × 2.1 mm, 2.0 μm particle size
Expand
View Pricing
50611-U
Ascentis
®
Express 90 Å ES-C18, 2.0 µm HPLC Column,
column L × I.D. 5 cm × 3.0 mm, particle size, std dev 2.0 μm
Expand
View Pricing
50617-U
Ascentis
®
Express 90 Å ES-C18, 2.0 µm HPLC Column,
column L × I.D. 10 cm × 3.0 mm, particle size, std dev 2.0 μm
Expand
View Pricing
50618-U
Ascentis
®
Express 90 Å ES-C18, 2.0 µm HPLC Column,
column L × I.D. 15 cm × 3.0 mm, particle size, std dev 2.0 μm
Expand
View Pricing