Skip to Content
CZ
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Neomycin trisulfate salt hydrate
Empirical Formula (Hill Notation):
C
23
H
46
N
6
O
13
· 3H
2
SO
4
· xH
2
O
CAS Number:
1405-10-3
Molecular Weight:
908.88 (anhydrous basis)
EC Number:
215-773-1
Products
Product Category
antibiotics (4)
Brand
Sigma-Aldrich (4)
Biological Source
Color
Feature
Formula Weight
Physical Form
Product Line
Activity Spectrum
Agency
Quality Segment
Application
Mode of Action
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
N6386
Neomycin trisulfate salt hydrate,
powder, BioReagent, suitable for cell culture
Expand
View Pricing
N1876
Neomycin trisulfate salt hydrate,
powder
Expand
View Pricing
N5285
Neomycin trisulfate salt hydrate,
meets USP testing specifications, powder
Expand
View Pricing
N3144
Neomycin trisulfate salt hydrate,
suitable for plant cell culture
Expand
View Pricing