Skip to Content
CZ
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
tert
-Butanol
Synonym(s):
2-Methyl-2-propanol,
tert
-Butyl alcohol, Trimethyl carbinol, tert-Butanol
Linear Formula:
(CH
3
)
3
COH
CAS Number:
75-65-0
Molecular Weight:
74.12
EC Number:
200-889-7
Products
Product Category
solvents (6)
Brand
Sigma-Aldrich (5)
Supelco (2)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Special Grade
ACS reagent (1)
Anhydrous (1)
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
360538
tert
-Butanol,
ACS reagent, ≥99.0%
Expand
View Pricing
471712
tert
-Butanol,
anhydrous, ≥99.5%
Expand
View Pricing
308250
tert
-Butanol,
suitable for HPLC, ≥99.5%
Expand
View Pricing
24127
tert
-Butanol,
≥99% (GC)
Expand
View Pricing
8.22264
tert
-Butanol,
EMPLURA
®
Expand
View Pricing
1.09629
tert
-Butanol,
for analysis EMSURE
®
ACS
Expand
View Pricing
B85927
tert
-Butanol,
TEBOL
®
99, ≥99.3%
Expand
View Pricing