Skip to Content
Merck
Tamoxifen citrate salt ≥99%

Tamoxifen citrate salt

Tamoxifen citrate salt ≥99%
Synonym(s):
(Z)-1-(p-Dimethylaminoethoxyphenyl)-1,2-diphenyl-1-butene, Tamoxifen Citrate
Empirical Formula (Hill Notation):
C26H29NO · xC6H8O7
CAS Number:
Molecular Weight:
371.51 (salt-free basis)

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-2 of 2