D-(+)-Trehalose dihydrate
Synonym(s):
α,α-Trehalose, α-D-Glucopyranosyl-α-D-glucopyranoside, D-(+)-Trehalose dihydrate, Trehalose, Dihydrate
Empirical Formula (Hill Notation):
C12H22O11 · 2H2O
CAS Number:
Molecular Weight:
378.33
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Product Line
Purity
Agency
Research Area
Tissue
Quality Segment
Application
Optical Isomer
Technique
Showing 1-7 of 7
Compare
Product Number
Description
Pricing