Skip to Content
Merck
Lamotrigine ≥98%, powder

Lamotrigine

Lamotrigine ≥98%, powder
Synonym(s):
6-(2,3-Dichlorophenyl)-1,2,4-triazine-3,5-diamine, GI 267119X
Empirical Formula (Hill Notation):
C9H7Cl2N5
CAS Number:
Molecular Weight:
256.09
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing