Skip to Content
Merck
Resveratrol ≥99% (HPLC)

Resveratrol

Resveratrol ≥99% (HPLC)
Synonym(s):
3,4′,5-Trihydroxy-trans-stilbene, 5-[(1E)-2-(4-Hydroxyphenyl)ethenyl]-1,3-benzenediol, Resveratrol
Empirical Formula (Hill Notation):
C14H12O3
CAS Number:
Molecular Weight:
228.24

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Tissue
Quality Segment
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing