Skip to Content
Merck
L-Glutamic acid ReagentPlus®, ≥99% (HPLC)

L-Glutamic acid

L-Glutamic acid ReagentPlus®, ≥99% (HPLC)
Synonym(s):
(S)-2-Aminopentanedioic acid, Glu, L-Glutamic acid, (S)-(+)-Glutamic acid
Linear Formula:
HO2CCH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
147.13

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Optical Isomer
Amino Acid
Technique