Skip to Content
Merck
2,2,4-Trimethylpentane suitable for HPLC, ≥99%

Isooctane

2,2,4-Trimethylpentane suitable for HPLC, ≥99%
Synonym(s):
2,2,4-Trimethylpentane, Isooctane
Linear Formula:
(CH3)2CHCH2C(CH3)3
CAS Number:
Molecular Weight:
114.23
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Special Grade
Technique