Xylenes
Synonym(s):
Xylene mixture of isomers
Linear Formula:
C6H4(CH3)2
CAS Number:
Molecular Weight:
106.17
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Agency
Chemical Composition
Quality Segment
Application
Special Grade
Showing 1-6 of 6