Key Documents
56033
Omnifit® Standard Bore Connector
Two-way valve connectors
About This Item
UNSPSC Code:
12000000
packaging
pkg of 2 units
manufacturer/tradename
Omnifit 1101
technique(s)
LPLC: suitable
bore
1.5 mm
compatibility
for use with 1/16 in. tubing
General description
Omnifit® Standard Bore Connector is very versatile in nature and is used for connecting various types of hardwall tubings. The standard bore connector has a manual valve operation on each end, thus the flow can be switched on or off as required. It is primarily used for fluid-line interconnection, sampling and mixing.
Application
Omnifit® Standard Bore Connector was used to construct the flow-injection system for amperometric determination of tacrine.
Features and Benefits
O-rings and PTFE cone included.
Legal Information
Omnifit is a registered trademark of Diba Industries
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Flow-Injection Amperometric Determination of Tacrine based on Ion Transfer across a Water-Plasticized Polymeric Membrane Interface.
Joaquin, A. C.
Sensors, 7, 1185-1192 (2007)
Modern Instrumental Techniques
Keith, A
Soil and Environmental Analysis, 1-700 (2003)