Cytokine, chemokine, and growth factor research plays a significant role in achieving a deeper understanding of the immune system and disease. This webcast will demonstrate an independent researcher’s use of a 48-plex panel designed to advance this aim.
The MILLIPLEX® Human Cytokine/Chemokine/Growth Factor Panel A simultaneously measures picogram levels of 48 immune factors using Luminex® xMAP® technology in small volumes of serum, plasma, or cell culture samples. This is the largest multiplex kit offered in the MILLIPLEX® portfolio, allowing researchers to save precious time and samples.
In this webcast, the speaker will examine the kit’s performance, compare key analytes to other assays, and share data generated with three very different disease samples: normal and sepsis samples, ovarian cancer samples, and autoimmune disease samples.
For Research Use Only. Not For Use In Diagnostic Procedures.
Speakers
Anna Lokshin
University of Pittsburgh Cancer Institute
Professor, Medicine, Pathology, Ob/Gyn; Director Luminex Core
Anna Lokshin received her B.S. (1973), M.S. (1975), and Ph.D. (1983) from University of Leningrad (Russia). She has been working in the area of cancer research at University of Pittsburgh for 28 years. Her work is focused on discovery of cancer biomarkers for early diagnosis and screening.
Protein biology
- Protein quantitation
Duration:50min
Language:English
Session 1:presented May 19, 2020