MTCT9045
MTC™ Bio Pestle for FlowTubes™ Strainer Caps
Sinonimo/i:
culture cap, strainer cap pestle
Confezionamento
pack of 100 ea (individually wrapped)
Produttore/marchio commerciale
MTC™ Bio T9045
Categorie correlate
Descrizione generale
Pestle for FlowTubes™, for use with 35µm strainer caps, sterile, individually wrapped, 100/pk
Caratteristiche e vantaggi
- Compatible with FACS (BD) instruments
- Available with 35µm strainer cap, standard cap, or no cap
- Clear polystyrene
- 5mL capacity
Note legali
FlowTubes is a trademark of MTC Bio
MTC is a trademark of MTC Bio