EP5424704000
Eppendorf® Rotor F-45-32-5-PCR
incl. lid
About This Item
Prodotti consigliati
Caratteristiche
to hold 4 x PCR strips
incl. lid
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Eppendorf® 5424704000
Parametri
15,000 max. RCF (18,615 xg)
tecniche
PCR: suitable
centrifugation: suitable
Compatibilità
for use with centrifuge models 5424/5424R
Categorie correlate
Note legali
Eppendorf is a registered trademark of Eppendorf AG