Key Documents
234S
Sigma-Aldrich
hCG Positive Control Slides
suitable for immunohistochemistry (formalin-fixed, paraffin-embedded sections)
About This Item
Codice UNSPSC:
12352200
NACRES:
NA.24
Confezionamento
pkg of 5 slides
Livello qualitativo
tecniche
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
Condizioni di spedizione
ambient
Temperatura di conservazione
no temp limit
Descrizione generale
hCG positive control slides are intended for use as positive controls for immunohistochemical (IHC) staining using an anti-hCG antibody.
Stato fisico
Paraffin embedded tissue section mounted on microscope slide
Altre note
For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com
Control slide products are manufactured from human biological samples (HBS). Purchaser shall not make any attempt to trace or identify donors from whom the HBS material was obtained. Purchaser shall use any data (including genetic information), generated with or by use of the HBS only in compliance with obligations of confidentiality and non-use equivalent with all applicable international and national legislation and guidelines on the protection of personal data and confidentiality. In addition to the use restrictions contained on the product label and/or product datasheet or other applicable use terms and conditions of sale, Purchaser agrees to use the HBS solely in accordance with the World Medical Association (WMA) Declaration of Helsinki adopted in 1964 as subsequently amended by WMA on Ethical Principles for Medical Research Involving Human Subjects.
Anticorpo
N° Catalogo
Descrizione
Determinazione del prezzo
Codice della classe di stoccaggio
11 - Combustible Solids
WGK 3