Documenti fondamentali
Z685305
Platinum crucible, with reinforced rim
capacity 50 mL
Sinonimo/i:
High-Temperature Crucible
About This Item
Codice UNSPSC:
41122200
NACRES:
NB.43
Produttore/marchio commerciale
Birmingham metals
Capacità
50 mL
Descrizione generale
Platinum labware has excellent resistance to acids such as acetic, hydrochloric, hydrofluoric, hydroiodic, nitric, selenic, and sulfuric, and to oxidation at high temperatures. These platinum crucibles can be cleaned by boiling in a suitable solvent, for the removal of substances which have alloyed with the surface, scour with alumina powder. Having a reinforced rim means that the rim of the crucible is thicker than the walls to make it stronger; this is where the crucible can be gripped if tongs are used during the handling process.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 9
1 of 9
Platinum combustion boat capacity 12 mL, with handle
Z685429
Platinum combustion boat
Platinum evaporating dish capacity 50 mL
Z562173
Platinum evaporating dish
Platinum crucible lid fits 30 mL crucible
Z562106
Platinum crucible lid
Platinum crucible lid fits 25 mL crucible
Z562084
Platinum crucible lid
Platinum crucible standard, capacity 15 mL
Z562041
Platinum crucible
Platinum crucible micro, capacity 5 mL
Z562238
Platinum crucible
Platinum crucible standard, capacity 10 mL
Z562025
Platinum crucible
Contatta l'Assistenza Tecnica