SYNS711754
Synthware™ high vacuum 90 degree stopcock with hollow plug
bore size 4 mm, plug size 17.35/50
Sinonimo/i:
Synthware S711754
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware S711754
Dimensioni foro
4 mm
Descrizione generale
Precision ground with corresponding numbers on the barrel and plug. Lower vacuum chamber is evacuated via a separate 2 mm hole in the plug which is not in the line of flow.
Note legali
Synthware is a trademark of Kemtech America Inc.