Small Molecule HPLC
Analysis of Small Molecules
A small molecule refers to a compound of low molecular weight (typically less than 900 daltons). Some common examples of small molecules include amino acids, lipids, sugars, fatty acids, alkaloids, and others.
Different methods are available for the separation of small molecules including High Performance liquid chromatography (HPLC), liquid chromatography (LC), gas chromatography (GC), thin-layer chromatography (TLC) and capillary electrophoresis (CE). Furthermore, the options for their identification include nuclear magnetic resonance spectroscopy (NMR) or mass spectrometry (MS). Liquid chromatography coupled with mass spectrometry (LC-MS) became a key technique for the identification of small molecules during recent years.
Getting the best possible results in the high-performance liquid chromatography (HPLC), UHPLC or LC-MS analysis of small molecules depends on the selection of the best suitable stationary phase and mobile phase conditions. The chemistry of the analyte is key for selecting the best suitable column chemistry. Other aspects such as speed, sample matrix, and number of compounds define the best suitable base material for the stationary phase.
Find Chromatograms
Related Technical Articles
- Chromolith® HPLC and UHPLC columns are made from monolithic silica with a bimodal pore structure using sol-gel technology.
- Review the effects of efficiency (plate number), retention, and selectivity factors on HPLC resolution. See how factors relate to particle and column characteristics via the HPLC resolution equation.
- Supel™ Carbon LC U/HPLC columns packed with porous graphitic carbon (PGC) particles, used for the HPLC, UHPLC, UPLC, and LC-MS separation of polar or charged compounds at high temperatures (up to 250 ˚), pressures up to 700 bar, and the mobile phase pH range of 1-14.
- Various measures and options for maximized LC-MS sensitivity and low limit of detection (LOD) are shown on this page. Each and every tip avoids contaminations causing signal suppression, adduct formation, elevated background noise and increased spectrum complexity.
- This study demonstrates the impact of various mobile phase modifiers on the separation; formate modifiers outperform acetate in terms of MS signals (or sensitivity) and chromatographic resolution.
- See All (186)
Related Protocols
- HPLC separation of 17 important cannabinoids including CBD, delta 9 THC and THCA. Read the application note
- Complete workflow for the comprehensive analysis of terpenes in cannabis
- Amino Acid Analysis is the suitable tool for precise determination of protein quantities but also provides detailed information regarding the relative amino.
- Following GB method for Vitamin A and Vitamin E as it is written with a 250 x 4.6 mm Ascentis® Express C30 column gives baseline resolution between the critical pair (gamma-, and beta-tocopherol; peaks 3 and 4).
- Alkylphenols are starting materials for the synthesis of alkylphenol ethoxylates, which are widely used as non ionic tensides, dispersive agents in paper and leather manufacturing, emulsifiers for pesticide formulations and as auxiliary agents for drilling and flotation.
- See All (181)
Find More Articles and Protocols
HPLC of Small Molecules
The HPLC analysis of small molecules most often is performed in reversed-phase separation mode. For separation of polar compounds hydrophilic interaction chromatography (HILIC) and normal-phase chromatography are suitable as well, with HILIC being the preferred method. For the separation of ionic compounds, ion-exchange separation modes and for inorganic anions or cations, ion chromatography can also be used.
The HPLC column is packed with either fully porous silica particles, superficially porous silica particles, polymeric particles or consists of a monolithic silica rod as the stationary phase. In addition, alumina oxide, zirconia particles and Carbon particles are used. The typical pore size of the stationary phase material for small molecule separation is in the range of 60 Å - 160 Å. For HPLC the typical particle size of the stationary phase ranges from 3 µm to 5 µm, for UHPLC smaller particles sizes, typically 2 µm or below, are used. Different column selectivities (modifications) can be attached to the stationary phase. A C18 alkyl chain is the most commonly used column chemistry in reversed phase (RP) chromatography. Nevertheless, other modifications, such as C30, C8, Phenyl, Pentaflourophenyl and a broad range of more polar modifications as well as modifications with ion-exchange or chiral properties enable the separation of almost all compounds soluble in liquids. The mobile phase for RP-HPLC typically consists of an aqueous buffer or water and water-miscible organic solvents like acetonitrile or methanol.
HPLC Sample Preparation
Complex and matrix-rich samples, such as food, beverages, cosmetics, biological samples and matrix-rich pharmaceutical formulations (e.g. cream, syrup) require efficient sample preparation protocols to remove unwanted components and selectively extract the analyte of interest. This is critical if a stationary phase with very small particle sizes as in UHPLC, where particles of 2 µm or below are used. Common sample preparation methods are Liquid-liquid extraction, Solid phase extraction (SPE) and for biological samples also protein precipitation beside filtration. In addition to the selective elution of the target compound and pre-concentration, the main purpose of sample preparation is to protect the HPLC stationary phase from clogging caused by the sample matrix. HPLC columns based on monolithic silica can tolerate matrix to a high extend and require much less sample preparation than particulate columns.
Derivatization
For some molecules derivatization is required, either before (pre-column) or after (post-column) the HPLC separation. The derivatization converts molecules into their derivatives for better sensitivity or chromatographic retention in HPLC. Chemical reagents, with desirable physical and chemical properties, are used for the required derivatization.
Highlights
Calculate the run-time and solvent-consumption savings when transferring method from HPLC to UHPLC.