- Prostaglandin E
Prostaglandin E
Gynecological endocrinology : the official journal of the International Society of Gynecological Endocrinology (2017-03-10)
Xiaman Huang, Haizhi Liu, Ruiman Li
PMID28277118
ABSTRACT
Integrin αvβ3 (ITG αvβ3) participates in the process of implantation between the embryo and the endometrium. This study investigated the effects of prostaglandin E Quantitative real-time reverse transcription PCR (qRT-PCR) and western blotting were used to detect the changes in mRNA and protein levels of ITG αvβ3 in RL95-2 cells after administering PGE PGE An appropriate concentration of PGE