Merck
Intracellular calcium release through IP
Neurobiology of disease (2017-07-16)
Ben C Orem, Nicolas Pelisch, Joshua Williams, Jacqueline M Nally, David P Stirling
ABSTRACT
Severed CNS axons often retract or dieback away from the injury site and fail to regenerate. The precise mechanisms underlying acute axonal dieback and secondary axonal degeneration remain poorly understood. Here we investigate the role of Ca