Merck
BhbZIP60 from Resurrection Plant
Frontiers in plant science (2017-03-14)
Bo Wang, Hong Du, Zhennan Zhang, Wenzhong Xu, Xin Deng
ABSTRACT
Adverse environmental conditions cause endoplasmic reticulum (ER) stress in plants. To mitigate ER stress damage, ER associated transcription factors and inositol-requiring enzyme-1 (IRE1)-mediated
Sigma-Aldrich
Lithium acetate, 99.95% trace metals basis