Merck
  • Evaluation of an intragastric challenge model for Shigella dysenteriae 1 in rhesus monkeys (Macaca mulatta) for the pre-clinical assessment of Shigella vaccine formulations.
Evaluation of an intragastric challenge model for Shigella dysenteriae 1 in rhesus monkeys (Macaca mulatta) for the pre-clinical assessment of Shigella vaccine formulations.
APMIS : acta pathologica, microbiologica, et immunologica Scandinavica (2013-09-14)
Dilara Islam, Nattaya Ruamsap, Patchariya Khantapura, Ajchara Aksomboon, Apichai Srijan, Boonchai Wongstitwilairoong, Ladaporn Bodhidatta, Montip Gettayacamin, Malabi M Venkatesan, Carl J Mason
ABSTRACT
Shigellosis is a worldwide disease, characterized by abdominal pain, fever, vomiting, and the passage of blood- and mucus-streaked stools. Rhesus monkeys and other primates are the only animals that are naturally susceptible to shigellosis. A suitable animal model is required for the pre-clinical evaluation of vaccines candidates. In this study, the minimal dose of Shigella dysenteriae1 1617 strain required to produce dysentery in four of five (80% attack rate) monkeys using an escalating dose range for three groups [2 × 10(8) , 2 × 10(9) and 2 × 10(10) colony forming unit (CFU)] was determined. In addition, the monkeys were re-infected. The identified optimal challenge dose was 2 × 10(9) CFU; this dose elicited 60% protection in monkeys when they were re-challenged with a one log higher dose (2 × 10(10) CFU). The challenge dose, 2 × 10(10) CFU, produced severe dysentery in all monkeys, with one monkey dying within 24 h, elicited 100% protection when re-challenged with the same dose. All monkeys exhibited immune responses. This study concludes that the rhesus monkey model closely mimics the disease and immune response seen in humans and is a suitable animal model for the pre-clinical evaluation of Shigella vaccine candidates. Prior infection with the 1617 strain can protect monkeys against subsequent re-challenges with homologous strains.
Sigma-Aldrich
Leucoberbelin Blue I, Dye content 65 %