Merck
  • Salicylic acid and some of its derivatives as antibacterial agents for viscose fabric.
Salicylic acid and some of its derivatives as antibacterial agents for viscose fabric.
International journal of biological macromolecules (2013-10-01)
A Kantouch, A Atef El-Sayed, M Salama, A Abou El-Kheir, S Mowafi
ABSTRACT
Salicylic acid and three of its derivatives were used to provide antibacterial properties to viscose fabrics. The four bactericides used were bonded to the viscose fabrics using epichlorohydrin or polymer binders. Optimization of the salicylic acid and its derivatives as well as the concentration of polymers was reported. The ability of the polymer binders to attract and bind the four bactericides was observed. The overall results show that the antibacterial reactivity of salicylic acid and its derivatives are in the following order 5-bromosalicylic acid>salicylic acid>5-chlorosalicylic acid>4-chlorosalicylic acid. Using epichlorohydrin as a binding agent, unfortunately, inhibits the bactericidal activity of the four bactericides. The FTIR study concludes that the reaction between salicylic acid as well as its derivatives with epichlorohydrin takes place through the phenolic group of the acids. The unexpected deterioration in the bactericidal properties of salicylic acid and its derivatives as a result of the treatment with epichlorohydrin could be due to the nature of interaction between the epichlorohydrin molecule and the acids molecules. PVP and PU show superior ability to sustain the four bactericides used even after 10 washing cycles.
Sigma-Aldrich
5-Bromosalicylic acid, technical grade, 90%
Pricing and availability is not currently available.