Merck
  • Dynein-dependent transport of spindle assembly checkpoint proteins off kinetochores toward spindle poles.
Dynein-dependent transport of spindle assembly checkpoint proteins off kinetochores toward spindle poles.
FEBS letters (2014-07-30)
Patrícia M A Silva, Rita M Reis, Victor M Bolanos-Garcia, Claudia Florindo, Álvaro A Tavares, Hassan Bousbaa
ABSTRACT
A predominant mechanism of spindle assembly checkpoint (SAC) silencing is dynein-mediated transport of certain kinetochore proteins along microtubules. There are still conflicting data as to which SAC proteins are dynein cargoes. Using two ATP reduction assays, we found that the core SAC proteins Mad1, Mad2, Bub1, BubR1, and Bub3 redistributed from attached kinetochores to spindle poles, in a dynein-dependent manner. This redistribution still occurred in metaphase-arrested cells, at a time when the SAC should be satisfied and silenced. Unexpectedly, we found that a pool of Hec1 and Mis12 also relocalizes to spindle poles, suggesting KMN components as additional dynein cargoes. The potential significance of these results for SAC silencing is discussed.
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Paraformaldehyde, powder, 95%
Sigma-Aldrich
Monoclonal Anti-Dynein (Intermediate Chain) antibody produced in mouse, clone 70.1, ascites fluid
Sigma-Aldrich
Nordihydroguaiaretic acid, ≥97.0% (HPLC)
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
DAPI, for nucleic acid staining