Merck
  • Interdigital tinea pedis (dermatophytosis simplex and complex) and treatment with ciclopirox 0.77% gel.
Interdigital tinea pedis (dermatophytosis simplex and complex) and treatment with ciclopirox 0.77% gel.
International journal of dermatology (2003-08-05)
Aditya K Gupta, Alayne R Skinner, Elizabeth A Cooper
ABSTRACT
The most common presentation of tinea pedis (athlete's foot) is that involving the interdigital spaces. Tinea pedis interdigitalis may present as asymptomatic dermatophytosis simplex or dermatophytosis complex, which is symptomatic, with secondary bacterial infection. In the dermatophytosis complex presentation there may be inflammation, maceration and odor, with bacterial involvement. Ciclopirox gel offers advantages in the treatment of tinea pedis, especially in the dermatophytosis complex presentation, with antifungal, antibacterial, and anti-inflammatory activity; furthermore, the gel formulation is fast drying, which is an advantage when the toe web area is moist.
Product Description
Ciclopirox, European Pharmacopoeia (EP) Reference Standard