Merck
  • Horizontal transfer and death of a fungal secondary metabolic gene cluster.
Horizontal transfer and death of a fungal secondary metabolic gene cluster.
Genome biology and evolution (2012-02-02)
Matthew A Campbell, Antonis Rokas, Jason C Slot
ABSTRACT
A cluster composed of four structural and two regulatory genes found in several species of the fungal genus Fusarium (class Sordariomycetes) is responsible for the production of the red pigment bikaverin. We discovered that the unrelated fungus Botrytis cinerea (class Leotiomycetes) contains a cluster of five genes that is highly similar in sequence and gene order to the Fusarium bikaverin cluster. Synteny conservation, nucleotide composition, and phylogenetic analyses of the cluster genes indicate that the B. cinerea cluster was acquired via horizontal transfer from a Fusarium donor. Upon or subsequent to the transfer, the B. cinerea gene cluster became inactivated; one of the four structural genes is missing, two others are pseudogenes, and the fourth structural gene shows an accelerated rate of nonsynonymous substitutions along the B. cinerea lineage, consistent with relaxation of selective constraints. Interestingly, the bik4 regulatory gene is still intact and presumably functional, whereas bik5, which is a pathway-specific regulator, also shows a mild but significant acceleration of evolutionary rate along the B. cinerea lineage. This selective preservation of the bik4 regulator suggests that its conservation is due to its likely involvement in other non-bikaverin-related biological processes in B. cinerea. Thus, in addition to novel metabolism, horizontal transfer of wholesale metabolic gene clusters might also be contributing novel regulation.
Sigma-Aldrich
Bikaverin, from Fusarium subglutinans, ≥98% (HPLC)