Merck
  • Chronic treatment with the thiazolidinedione, MCC-555, is associated with reductions in nitric oxide synthase activity and beta-cell apoptosis in the pancreas of the Zucker Diabetic Fatty rat.
Chronic treatment with the thiazolidinedione, MCC-555, is associated with reductions in nitric oxide synthase activity and beta-cell apoptosis in the pancreas of the Zucker Diabetic Fatty rat.
International journal of experimental pathology (2003-06-13)
Lucy C Pickavance, Peter S Widdowson, John R Foster, Gareth Williams, John P H Wilding
ABSTRACT
The Zucker Diabetic Fatty (ZDF) rat is a model of impaired insulin sensitivity arising from hyperphagia owing to a mutation in the leptin receptor. In time, young ZDF rats, which are not initially diabetic, develop impaired pancreatic beta-cell function leading to apoptotic cell death. This results in an inability to fully compensate for the reduction in insulin sensitivity with hypersecretion of insulin. Young, pre-diabetic ZDF rats were treated, over a 4-week period, with the thiazolidinedione compound MCC-555, and the islet morphology studied in comparison to ZDF rats not given MCC-555. In particular, changes in the apoptotic incidence, as measured using TUNEL staining to localize apoptotic cells, were studied over the 4-week period. Changes in the induction of nitric oxide synthase and in the accumulation of nitrate/nitrite within the pancreas were also studied during the time course of administration of MCC-555. The study has demonstrated that the administration of MCC-555 significantly decreases the apoptotic incidence in the islets of Langerhans of pre-diabetic ZDF rats given the compound, as compared to those not given MCC-555, as well as decreasing the accumulation of nitrate/nitrite within the pancreas.
Sigma-Aldrich
MCC-555, ≥98% (HPLC)
Pricing and availability is not currently available.