Merck
Environmental noise degrades hippocampus-related learning and memory.
Proceedings of the National Academy of Sciences of the United States of America (2020-11-25)
Yifan Zhang, Min Zhu, Yutian Sun, Binliang Tang, Guimin Zhang, Pengying An, Yuan Cheng, Ye Shan, Michael M Merzenich, Xiaoming Zhou
ABSTRACT
The neural mechanisms underlying the impacts of noise on nonauditory function, particularly learning and memory, remain largely unknown. Here, we demonstrate that rats exposed postnatally (between postnatal days 9 and 56) to structured noise delivered at a sound pressure level of ∼65 dB displayed significantly degraded hippocampus-related learning and memory abilities. Noise exposure also suppressed the induction of hippocampal long-term potentiation (LTP). In parallel, the total or phosphorylated levels of certain LTP-related key signaling molecules in the synapses of the hippocampus were down-regulated. However, no significant changes in stress-related processes were found for the noise-exposed rats. These results in a rodent model indicate that even moderate-level noise with little effect on stress status can substantially impair hippocampus-related learning and memory by altering the plasticity of synaptic transmission. They support the importance of more thoroughly defining the unappreciated hazards of moderately loud noise in modern human environments.