Merck
  • ASF1a inhibition induces p53-dependent growth arrest and senescence of cancer cells.
ASF1a inhibition induces p53-dependent growth arrest and senescence of cancer cells.
Cell death & disease (2019-01-30)
Yujiao Wu, Xidan Li, Jingya Yu, Magnus Björkholm, Dawei Xu
ABSTRACT
Anti-silencing function 1a (ASF1a) is a histone H3-H4 chaperone isoform involved in chromatin assembling and transcription regulation. Recently, ASF1a has been shown to be up-regulated in certain human malignancies and required for the expression of telomerase reverse transcriptase (TERT), a factor essential for the immortal phenotype of cancer cells; however, its role in oncogenesis remains poorly defined. In the present study, we determine whether ASF1a is required for the unlimited proliferation of cancer cells, a key cancer hallmark. Elevated ASF1a mRNA expression was observed in hepatocellular carcinoma (HCC) tumors. The overexpression of ASF1a was similarly found in 20 cancer types contained in TCGA and GTEx datasets. ASF1a knockdown led to growth arrest and senescence of wild-type (wt) p53-carrying HCC and prostate cancer cells. Cellular senescence mediated by ASF1a inhibition resulted from the robust up-regulation of p53 and p21cip1 expression, but without detectable changes in TERT expression. p53 inhibition attenuated p21cip1 induction caused by ASF1a depletion. Mechanistically, ASF1a-knocked down cells displayed widespread DNA damage. The TCGA dataset analysis revealed a negative correlation between ASF1a and p21cip1 expression in multiple types of primary tumors, including HCC, prostate, gastric, and breast cancer. Higher ASF1a and lower p21cip1 expression predicted a poor outcome in patients with HCC. Our results reveal that ASF1a overexpression is widespread in human malignancies and is required for the infinite proliferation of cancer cells, whereas its inhibition induces DNA damage and subsequent up-regulation of p53-p21cip1 expression, thereby triggering cellular senescence. Thus, ASF1a may serve as a potential target in cancer therapy.
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys9) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Cellular Senescence Assay, Cellular Senescence Assay Kits provide all the reagents required to efficiently detect SA-β-gal activity at pH 6.0 in cultured cells & tissue sections.