Merck
  • Histone H3k9 and H3k27 Acetylation Regulates IL-4/STAT6-Mediated Igε Transcription in B Lymphocytes.
Histone H3k9 and H3k27 Acetylation Regulates IL-4/STAT6-Mediated Igε Transcription in B Lymphocytes.
Anatomical record (Hoboken, N.J. : 2007) (2015-05-09)
Xiao Fu, Xinting Wang, Zhongchao Duan, Chunyan Zhang, Xue Fu, Jie Yang, Xin Liu, Jinyan He
ABSTRACT
IL-4 activates STAT6 and causes the subsequent up-regulation of Ig heavy chain germline Igε via chromatin remodeling involved in B lymphocytes development. STAT6 acts as a molecular switch to regulate the higher-order chromatin remodeling via dynamically orchestrating co-activators (CBP/Tudor-SN) and co-repressors (HDAC1/PSF). Here, we demonstrated that STAT6/Tudor-SN/PSF form a complex, balancing the acetylation and deacetylation states to co-regulate IL-4/STAT6 gene transcription. In addition, we confirmed that IL-4 treatment increased the HATs activity in Ramos cells. As "active" markers, the expression of H3K9ac and H3K27ac increased after treatment with IL-4. However, transcriptional repressors such as H3K9me3 and H3K27me3 decreased in response to IL-4 stimulation. Moreover, IL-4 treatment enhanced H3 acetylation at the Igε promoter regions. Our results revealed that the Igε gene transcription is regulated by histone modifications in the IL-4/STAT6 pathway. The study will provide novel insights into the pathogenesis of allergic diseases.
Sigma-Aldrich
Anti-acetyl-Histone H3 (Lys 4) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Monoclonal Anti-PSF antibody produced in mouse, clone B92, purified from hybridoma cell culture