Metformin Related Compound A
Synonym(s):
Dicyandiamide, Cyanoguanidine, Dicyanodiamide
Linear Formula:
NH2C(=NH)NHCN
CAS Number:
Molecular Weight:
84.08
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing