Skip to Content
Merck
Sorbitol European Pharmacopoeia (EP) Reference Standard

Sorbitol

Sorbitol European Pharmacopoeia (EP) Reference Standard
Synonym(s):
D-Sorbitol, D-Glucitol
Empirical Formula (Hill Notation):
C6H14O6
CAS Number:
Molecular Weight:
182.17

Products

Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Optical Isomer
API Family
Special Grade
Technique