Ammonium ZSM-5 Zeolite
Synonym(s):
Zeolite, Synthetic zeolite, Dioxosilane, Oxo(oxoalumanyloxy)alumane
Linear Formula:
Na4[(AlO2)4(SiO2)4] · 9H2O
CAS Number:
Molecular Weight:
730.36
Products
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Showing 1-6 of 6
Compare
Product Number
Description
Pricing