Skip to Content
CH
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sulfuric acid
Synonym(s):
Sulfuric acid solution
Linear Formula:
H
2
SO
4
CAS Number:
7664-93-9
Molecular Weight:
98.08
Products
Brand
Sigma-Aldrich (1)
Supelco (12)
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Package Size
Reaction Type
Special Grade
Puriss (1)
Puriss P.A. (1)
Technique
Filter Products
Showing 1-16 of 16
Compare
Product Number
Description
Pricing
1.09072
Sulfuric acid,
c(H
2
SO
4
) = 0.5 mol/l (1 N), Titripur
®
, reag. Ph. Eur., reag. USP
Expand
View Pricing
1.09074
Sulfuric acid,
c(H
2
SO
4
) = 0.05 mol/l (0.1 N), Titripur
®
, reag. USP
Expand
View Pricing
68279
Sulfuric acid,
0.1 M H
2
SO
4
in water (0.2N), eluent concentrate for IC
Expand
View Pricing
1.60313
Sulfuric acid,
c(H
2
SO
4
) = 1 mol/l (2 N), Titripur
®
Expand
View Pricing
1.00716
Sulfuric acid,
for analysis EMSURE
®
Expand
View Pricing
1.09981
Sulfuric acid,
for 1000 ml, c(H₂SO₄) = 0.5 mol/l (1 N) Titrisol
®
Expand
View Pricing
1.60315
Sulfuric acid,
5 M H
2
SO
4
(10 N), Titripur
®
Expand
View Pricing
84736
Sulfuric acid,
puriss. p.a., ≥25% (T)
Expand
View Pricing
1.09073
Sulfuric acid,
c(H
2
SO
4
)=0,25MOL/L (0.5N) c(H
2
SO
4
) = 0.25 mol/l (0.5 N), Titripur
®
Expand
View Pricing
1.09984
Sulfuric acid,
for 1000 ml, c(H₂SO₄) = 0.05 mol/l (0.1 N) Titrisol
®
Expand
View Pricing
4.80364
Sulfuric acid,
c(H
2
SO
4
) = 2.5 mol/l (5 N), Titripur
®
Expand
View Pricing
1.09286
Sulfuric acid,
for determination of gas metabolism acc. to Knipping
Expand
View Pricing
1.60314
Sulfuric acid,
c(H
2
SO
4
) = 0.005 mol/l (0.01 N), Titripur
®
, reag. USP
Expand
View Pricing
1.09912
Sulfuric acid,
for the preparation of dilute volumetric solutions or for direct use c(H₂SO₄) = 2.5 mol/l (5 N) Combi-Titrisol
Expand
View Pricing
4.80531
Sulfuric acid,
for the determination of fat in cheese (d 1.52) EMSURE
®
Expand
View Pricing
1.09982
Sulfuric acid,
for 1000 ml, c(H₂SO₄) = 0.005 mol/l (0.01 N) Titrisol
®
Expand
View Pricing