Skip to Content
CH
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Molecular Sieve Adsorbent
Products
Product Category
sieves (5)
Brand
Supelco (5)
Physical Form
Agency
Quality Segment
Package Size
Pore Size (Å)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
20300
Molecular Sieve Adsorbent,
matrix Molecular Sieve 5A, 30-40 mesh, bottle of 50 g
Expand
View Pricing
20302
Molecular Sieve Adsorbent,
matrix Molecular Sieve 5A, 60-80 mesh, bottle of 50 g
Expand
View Pricing
20305
Molecular Sieve Adsorbent,
matrix Molecular Sieve 13X, 60-80 mesh, bottle of 50 g
Expand
View Pricing
20301
Molecular Sieve Adsorbent,
matrix Molecular Sieve 5A, 45-60 mesh, bottle of 50 g
Expand
View Pricing
20307
Molecular Sieve Adsorbent,
matrix Molecular Sieve 13X, 100-120 mesh, bottle of 50 g
Expand
View Pricing