Skip to Content
CH
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Estapor
®
Carboxylated Colored Microspheres
Products
Brand
Sigma-Aldrich (4)
Color
Feature
Physical Form
Quality Segment
Functional Group
Package Size
Particle Size (µm)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
FR180380637
Estapor
®
Carboxylated Colored Microspheres,
K1-030 Red Int COOH PS 5%,
Red Colored
Carboxylated microspheres
300nm
, Lateral Flow Immunoassay (LFIA), IVD
Expand
View Pricing
FR180380642
Estapor
®
Carboxylated Colored Microspheres,
K1-030 Blue Int COOH PS 5%
Blue Colored
Carboxylated microspheres
300nm
, Lateral Flow Immunoassay
Expand
View Pricing
FR180380638
Estapor
®
Carboxylated Colored Microspheres,
K1-040 Red Int COOH PS 5%,
Red Colored
Carboxylated microspheres
400nm
, Lateral Flow Immunoassay, LFIA, IVD
Expand
View Pricing
FR180380641
Estapor
®
Carboxylated Colored Microspheres,
K1-040 Blue Int COOH PS 5%
Blue Colored
Carboxylated microspheres
400nm
, Lateral Flow Immunoassay
Expand
View Pricing